Acetic Acid, 99.8%, for analysis
Acetic Acid, 99.8%, for analysis
Acetic Acid, 99.8%, for analysis
Acetic Acid, 99.8%, for analysis
Thermo Scientific Chemicals
Acetic Acid, 99.8%, for analysis
CAS: 64-19-7 | C2H4O2 | 60.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
250 mL
2.5 L
20 L
Packaging:
View all
Glass Bottle
Catalog number FSA222140010
Price (MYR)
490.00
BOT
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
490.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAcetic acid
Name NoteFor Analysis
CAS64-19-7
Melting Point16.0°C to 16.5°C
Boiling Point117.0°C to 118.0°C
View more
For analysis
RUO – Research Use Only