Methyl 3-bromopropionate, 97%
Methyl 3-bromopropionate, 97%
Methyl 3-bromopropionate, 97%
Methyl 3-bromopropionate, 97%
Thermo Scientific Chemicals
Methyl 3-bromopropionate, 97%
CAS: 3395-91-3 | C4H7BrO2 | 167.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
Packaging:
Glass bottle
Catalog number FSA221790250
Price (MYR)
726.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
726.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl 3-bromopropionate
Name Note97%
CAS3395-91-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: