Aniline, 99.5%, extra pure
Aniline, 99.5%, extra pure
Aniline, 99.5%, extra pure
Aniline, 99.5%, extra pure
Thermo Scientific Chemicals
Aniline, 99.5%, extra pure
CAS: 62-53-3 | C6H7N | 93.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA221732500
Price (MYR)
265.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
265.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAniline
CAS62-53-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing genetic defects.
Toxic in contact with skin.
Toxic if swallowed.
Causes serious eye dam
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF