Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Tetraethylammonium tetrafluoroborate, 99%
Thermo Scientific Chemicals
Tetraethylammonium tetrafluoroborate, 99%
CAS: 429-06-1 | C8H20BF4N | 217.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA221615000
Price (MYR)
7,286.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,286.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetraethylammonium tetrafluoroborate
CAS429-06-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
View more
Phase transfer catalyst
RUO – Research Use Only