sec-Butanol, 99+%, for analysis
sec-Butanol, 99+%, for analysis
sec-Butanol, 99+%, for analysis
sec-Butanol, 99+%, for analysis
Thermo Scientific Chemicals
sec-Butanol, 99+%, for analysis
CAS: 78-92-2 | C4H10O | 74.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA220290025
Price (MYR)
850.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
850.00
EA
Add to cart
Specifications
Chemical Name or Materialsec-Butanol
Melting Point-115.0°C
Name NoteFor analysis
CAS78-92-2
Health Hazard 1GHS Signal Word: Warning