(±)-Propylene oxide, 99.5%, extra pure
(±)-Propylene oxide, 99.5%, extra pure
(±)-Propylene oxide, 99.5%, extra pure
(±)-Propylene oxide, 99.5%, extra pure
Thermo Scientific Chemicals
(±)-Propylene oxide, 99.5%, extra pure
CAS: 75-56-9 | C3H6O | 58.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass bottle
Catalog number FSA220160010
Price (MYR)
397.00
EA
Add to cart
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
397.00
EA
Add to cart
Specifications
Chemical Name or Material(±)-Propylene oxide
Name NoteExtra Pure
CAS75-56-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Extremely flammable liquid and vapor.
Harmful if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
May cause cancer.