Hydroquinone, 99.5%
Hydroquinone, 99.5%
Hydroquinone, 99.5%
Thermo Scientific Chemicals
Hydroquinone, 99.5%
CAS: 123-31-9 | C6H6O2 | 110.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 g
50 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA219932500
Price (MYR)
299.00
EA
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
299.00
EA
Add to cart
Specifications
Chemical Name or MaterialHydroquinone
Name Notep.a.
CAS123-31-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye damage.
Suspected of causing genetic defects.
May cause an allergic skin reaction.
Suspected of causing cancer.
Harmful if swallowed.
Very toxic to aquatic life.