Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Triethylamine, 99.7%, extra pure
Thermo Scientific Chemicals
Triethylamine, 99.7%, extra pure
CAS: 121-44-8 | C6H15N | 101.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 kg
Catalog number FSA219510050
Price (MYR)
234.00
BOT
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (MYR)
234.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTriethylamine
Name Note99.7%
CAS121-44-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.