o-Phenylenediamine dihydrochloride, 99+%
o-Phenylenediamine dihydrochloride, 99+%
o-Phenylenediamine dihydrochloride, 99+%
o-Phenylenediamine dihydrochloride, 99+%
Thermo Scientific Chemicals
o-Phenylenediamine dihydrochloride, 99+%
CAS: 615-28-1 | C6H8N2·2HCl | 181.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA218481000
Price (MYR)
1,478.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,478.00
BOT
Add to cart
Specifications
Chemical Name or Materialo-Phenylenediamine dihydrochloride
Name Note99+%
CAS615-28-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger