Phenyl chloroformate, 99%
Phenyl chloroformate, 99%
Phenyl chloroformate, 99%
Phenyl chloroformate, 99%
Thermo Scientific Chemicals
Phenyl chloroformate, 99%
CAS: 1885-14-9 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Catalog number FSA218212500
Price (MYR)
770.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
770.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPhenyl chloroformate
CAS1885-14-9
Health Hazard 1GHS Signal Word: Danger
Melting Point-28.0°C
Density1.2440g/mL
View more
Coupling reagent
RUO – Research Use Only