(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee
(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee
(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee
(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee
Thermo Scientific Chemicals
(2R,3R)-(-)-2,3-Butanediol, 98+%, 99+% ee
CAS: 24347-58-8 | C4H10O2 | 90.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA217980050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(2R, 3R)-(-)-2, 3-Butanediol
Name Note≥99% ee
CAS24347-58-8
CAS Min %98.0
CAS Max %100.0