3-(Trimethoxysilyl)propyl methacrylate, 98%
3-(Trimethoxysilyl)propyl methacrylate, 98%
3-(Trimethoxysilyl)propyl methacrylate, 98%
3-(Trimethoxysilyl)propyl methacrylate, 98%
Thermo Scientific Chemicals
3-(Trimethoxysilyl)propyl methacrylate, 98%
CAS: 2530-85-0 | C10H20O5Si | 248.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA216555000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material3-(Trimethoxysilyl)propyl methacrylate
CAS2530-85-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0