Vinyltrimethoxysilane, 98%
Vinyltrimethoxysilane, 98%
Vinyltrimethoxysilane, 98%
Vinyltrimethoxysilane, 98%
Thermo Scientific Chemicals
Vinyltrimethoxysilane, 98%
CAS: 2768-02-7 | C5H12O3Si | 148.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA216521000
Price (MYR)
390.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
390.00
BOT
Add to cart
Specifications
Chemical Name or MaterialVinyltrimethoxysilane
CAS2768-02-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest