2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
Thermo Scientific Chemicals
2-(Dimethylamino)ethyl methacrylate, 99%, stabilized
CAS: 2867-47-2 | C8H15NO2 | 157.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA215842500
Price (MYR)
695.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
695.00
BOT
Add to cart
Specifications
Chemical Name or Material2-(Dimethylamino)ethyl methacrylate
Name NoteStabilized
CAS2867-47-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning