(2-Chloroethyl)trimethylammonium chloride, 98%
(2-Chloroethyl)trimethylammonium chloride, 98%
(2-Chloroethyl)trimethylammonium chloride, 98%
(2-Chloroethyl)trimethylammonium chloride, 98%
Thermo Scientific Chemicals
(2-Chloroethyl)trimethylammonium chloride, 98%
CAS: 999-81-5 | C5H13Cl2N | 158.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Catalog number FSA215420250
Price (MYR)
369.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
369.00
BOT
Add to cart
Specifications
Chemical Name or Material(2-Chloroethyl)trimethylammonium chloride
Name Note98%
CAS999-81-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
View more
Phase transfer catalyst
RUO – Research Use Only