Spermidine trihydrochloride, 99+%
Spermidine trihydrochloride, 99+%
Spermidine trihydrochloride, 99+%
Spermidine trihydrochloride, 99+%
Thermo Scientific Chemicals
Spermidine trihydrochloride, 99+%
CAS: 334-50-9 | C7H22Cl3N3 | 254.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA215100050
Price (MYR)
797.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
797.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSpermidine trihydrochloride
Name Note99+%
CAS334-50-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement