Serotonin hydrochloride, 97+%
Serotonin hydrochloride, 97+%
Serotonin hydrochloride, 97+%
Serotonin hydrochloride, 97+%
Thermo Scientific Chemicals
Serotonin hydrochloride, 97+%
CAS: 153-98-0 | C10H12N2O·HCl | 212.68 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mg
100 mg
Packaging:
Glass bottle
Catalog number FSA215025000
Price (MYR)
1,792.00
BOT
Add to cart
Quantity:
500 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,792.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSerotonin hydrochloride
Name Note99%
CAS153-98-0
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement