HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
HEPES sodium salt, 99%
Thermo Scientific Chemicals
HEPES sodium salt, 99%
CAS: 75277-39-3 | C8H17N2NaO4S | 260.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA215000010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Absorbance(0.1 M aq. soln.), (0.1 M aq. soln.) at 0nm, 0.02 at 280nm, 0.04 at 260nm
Chemical Name or MaterialHEPES sodium salt
Absorbance Conditions(0.1M aq. soln.)
Name Note99%
CAS75277-39-3
View more
Biological buffer
RUO – Research Use Only