Triisopropylsilane, 98%
Triisopropylsilane, 98%
Triisopropylsilane, 98%
Triisopropylsilane, 98%
Thermo Scientific Chemicals
Triisopropylsilane, 98%
CAS: 6485-79-6 | C9H22Si | 158.36 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA214922500
Price (MYR)
2,124.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,124.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriisopropylsilane
Name Note98%
CAS6485-79-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Flammable liquid and vapor.
View more
Reduction reagent
RUO – Research Use Only