Vanadyl(IV) sulfate hydrate, 17-23% V
Vanadyl(IV) sulfate hydrate, 17-23% V
Vanadyl(IV) sulfate hydrate, 17-23% V
Vanadyl(IV) sulfate hydrate, 17-23% V
Thermo Scientific Chemicals
Vanadyl(IV) sulfate hydrate, 17-23% V
CAS: 12439-96-2 | O5SV·xH2O | 163 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA214841000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialVanadyl(IV) sulfate hydrate
Name Note17-23% V
CAS12439-96-2
ColorWhite to Yellow
Health Hazard 1GHS Signal Word: Warning