Molecular sieves 3A, powder <50 micron
Molecular sieves 3A, powder &lt;50 micron
Molecular sieves 3A, powder &lt;50 micron
Molecular sieves 3A, powder &lt;50 micron
Thermo Scientific Chemicals
Molecular sieves 3A, powder <50 micron
CAS: 308080-99-1 | AlNaO6Si2 | 202.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA214795000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMolecular sieves 3A
ColorUndesignated
PackagingGlass bottle
Particle Size500 Micron Max.
Residual Water< 1.5% (550°C, 2 hrs.)
View more
Drying agent
RUO – Research Use Only