(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%
(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%
(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%
(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%
Thermo Scientific Chemicals
(alpha,alpha,alpha-Trifluoro-p-tolyl)acetic acid, 98%
CAS: 32857-62-8 | C9H7F3O2 | 204.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA214370010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(α, α, α-Trifluoro-p-tolyl)acetic acid
CAS32857-62-8
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0