[Bis(trifluoroacetoxy)iodo]benzene, 98%
[Bis(trifluoroacetoxy)iodo]benzene, 98%
[Bis(trifluoroacetoxy)iodo]benzene, 98%
[Bis(trifluoroacetoxy)iodo]benzene, 98%
Thermo Scientific Chemicals
[Bis(trifluoroacetoxy)iodo]benzene, 98%
CAS: 2712-78-9 | C10H5F6IO4 | 430.03 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA21367010010 gGlass bottle
Catalog number FSA213670100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material[Bis(trifluoroacetoxy)iodo]benzene
Name Note98%
CAS2712-78-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
oxidation reagent
RUO – Research Use Only