Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Cobalt(II) nitrate hexahydrate, 99%, pure
Thermo Scientific Chemicals
Cobalt(II) nitrate hexahydrate, 99%, pure
CAS: 10026-22-9 | CoH12N2O12 | 291.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA213095000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialCobalt(II) nitrate hexahydrate
Melting Point55.0°C
Name Note99%
CAS10026-22-9
ColorBrown-Red to Pink