Zinc bromide, 99.999%, (trace metal basis), extra pure
Zinc bromide, 99.999%, (trace metal basis), extra pure
Zinc bromide, 99.999%, (trace metal basis), extra pure
Zinc bromide, 99.999%, (trace metal basis), extra pure
Thermo Scientific Chemicals
Zinc bromide, 99.999%, (trace metal basis), extra pure
CAS: 7699-45-8 | Br2Zn | 225.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
Packaging:
Glass bottle
Catalog number FSA212770100
Price (MYR)
915.00
BOT
Add to cart
Quantity:
10 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
915.00
BOT
Add to cart
Specifications
Boiling Point697.0°C
Chemical Name or MaterialZinc bromide
Melting Point394.0°C
Name Note99.999%
CAS7699-45-8
View more
For ultra trace analysis
RUO – Research Use Only