Ammonium cerium(IV) nitrate, pure
Ammonium cerium(IV) nitrate, pure
Ammonium cerium(IV) nitrate, pure
Ammonium cerium(IV) nitrate, pure
Thermo Scientific Chemicals
Ammonium cerium(IV) nitrate, pure
Oxidation reagent | CAS: 16774-21-3 | H8CeN8O18 | 548.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Catalog number FSA212360010
Price (MYR)
798.00
EA
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (MYR)
798.00
EA
Add to cart
Specifications
Chemical Name or MaterialAmmonium cerium(IV) nitrate
Melting Point107.0°C to 108.0°C
CAS16774-21-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May intensify fire; oxidizer.
May be corrosive to metals.