Indigo, synthetic
Indigo, synthetic
Indigo, synthetic
Thermo Scientific Chemicals
Indigo, synthetic
CAS: 482-89-3 | C16H10N2O2 | 262.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA212130250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIndigo
Name Notesynthetic
CAS482-89-3
RTECS NumberDU2988400
EINECS Number207-586-9
View more
Indicator
RUO – Research Use Only