Dimethylethylhexadecylammonium bromide, 99%
Dimethylethylhexadecylammonium bromide, 99%
Dimethylethylhexadecylammonium bromide, 99%
Dimethylethylhexadecylammonium bromide, 99%
Dimethylethylhexadecylammonium bromide, 99%
CAS: 124-03-8 | C20H44BrN | 378.483 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA211870050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDimethylethylhexadecylammonium bromide
ColorWhite
Name Note99%
CAS124-03-8
Health Hazard 1HYGROSCOPIC
View more
Phase transfer catalyst
RUO – Research Use Only