Lithium, 99+%, granular, dry
Lithium, 99+%, granular, dry
Lithium, 99+%, granular, dry
Lithium, 99+%, granular, dry
Thermo Scientific Chemicals
Lithium, 99+%, granular, dry
CAS: 7439-93-2 | Li | 6.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Catalog number FSA211442500
Price (MYR)
4,618.00
BOT
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
4,618.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium
Name Note99+%
CAS7439-93-2
Melting Point180.0°C
Boiling Point1340.0°C