Tetrapropylammonium bromide, 98%
Tetrapropylammonium bromide, 98%
Tetrapropylammonium bromide, 98%
Tetrapropylammonium bromide, 98%
Thermo Scientific Chemicals
Tetrapropylammonium bromide, 98%
CAS: 1941-30-6 | C12H28BrN | 266.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA209375000
Price (MYR)
1,039.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,039.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTetrapropylammonium bromide
Name Note98%
CAS1941-30-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
Phase transfer catalyst
RUO – Research Use Only