2-Methylbenzofuran, 96%
2-Methylbenzofuran, 96%
2-Methylbenzofuran, 96%
2-Methylbenzofuran, 96%
Thermo Scientific Chemicals
2-Methylbenzofuran, 96%
CAS: 4265-25-2 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA208400010
Price (MYR)
932.00
EA
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
932.00
EA
Add to cart
Specifications
Chemical Name or Material2-Methylbenzofuran
CAS4265-25-2
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0