Thiocarbohydrazide, 98%
Thiocarbohydrazide, 98%
Thiocarbohydrazide, 98%
Thiocarbohydrazide, 98%
Thermo Scientific Chemicals
Thiocarbohydrazide, 98%
CAS: 2231-57-4 | CH6N4S | 106.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA207530050
Price (MYR)
299.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
299.00
EA
Add to cart
Specifications
Chemical Name or MaterialThiocarbohydrazide
Name Note98%
CAS2231-57-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Fatal if inhaled.