Tetramethylammonium hydroxide pentahydrate, 97%
Tetramethylammonium hydroxide pentahydrate, 97%
Tetramethylammonium hydroxide pentahydrate, 97%
Tetramethylammonium hydroxide pentahydrate, 97%
Thermo Scientific Chemicals
Tetramethylammonium hydroxide pentahydrate, 97%
CAS: 10424-65-4 | C4H23NO6 | 181.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA207525000
Price (MYR)
4,324.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
4,324.00
EA
Add to cart
Specifications
Chemical Name or MaterialTetramethylammonium hydroxide pentahydrate
CAS10424-65-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Toxic to aquatic life w
Health Hazard 3GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
View more
Phase transfer catalyst
RUO – Research Use Only