TES, 99%, for biochemistry
TES, 99%, for biochemistry
TES, 99%, for biochemistry
TES, 99%, for biochemistry
Thermo Scientific Chemicals
TES, 99%, for biochemistry
CAS: 7365-44-8 | C6H15NO6S | 229.247 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA207511000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Absorbance(0.1M aq. soln.), (0.1M aq. soln.) at 0nm, 0.02 max. at 280nm, 0.05 max. at 260nm
Chemical Name or MaterialTES
Absorbance Conditions(0.1M aq. soln.)
Name NoteFor biochemistry
CAS7365-44-8
View more
Biological buffer
RUO – Research Use Only