Ethyl stearate, 97%
Ethyl stearate, 97%
Ethyl stearate, 97%
Thermo Scientific Chemicals
Ethyl stearate, 97%
CAS: 111-61-5 | C20H40O2 | 312.52 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA207480250
Price (MYR)
844.00
EA
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
844.00
EA
Add to cart
Specifications
Chemical Name or MaterialEthyl stearate
Name Note97%
CAS111-61-5
Melting Point30°C to 39°C