DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
DL-Propranolol hydrochloride, 99%
Thermo Scientific Chemicals
DL-Propranolol hydrochloride, 99%
CAS: 318-98-9 | C16H21NO2·HCl | 295.8 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA207320050
Price (MYR)
524.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
524.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDL-Propranolol hydrochloride
CAS318-98-9
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful if swallowed.
Health Hazard 3GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.