Zirconium(IV) chloride, 98%, anhydrous
Zirconium(IV) chloride, 98%, anhydrous
Zirconium(IV) chloride, 98%, anhydrous
Zirconium(IV) chloride, 98%, anhydrous
Thermo Scientific Chemicals
Zirconium(IV) chloride, 98%, anhydrous
CAS: 10026-11-6 | Cl4Zr | 233.03 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA206415000
Price (MYR)
1,586.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,586.00
BOT
Add to cart
Specifications
Chemical Name or MaterialZirconium(IV) chloride
Melting Point437.0°C
Name Note98%
CAS10026-11-6