Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Molybdenum(VI) oxide, 99+%
Thermo Scientific Chemicals
Molybdenum(VI) oxide, 99+%
CAS: 1313-27-5 | MoO3 | 143.95 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA206365000
Price (MYR)
1,391.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,391.00
BOT
Add to cart
Specifications
Boiling Point1155.0°C
Chemical Name or MaterialMolybdenum(VI) oxide
Melting Point795.0°C
Name Note99+%
CAS1313-27-5