Heneicosanoic acid, 99%
Heneicosanoic acid, 99%
Heneicosanoic acid, 99%
Heneicosanoic acid, 99%
Thermo Scientific Chemicals
Heneicosanoic acid, 99%
CAS: 2363-71-5 | C21H42O2 | 326.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Packaging:
Glass bottle
Catalog number FSA204691000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHeneicosanoic acid
Name Note99%
CAS2363-71-5
Melting Point74.0°C to 76.0°C