2,6-Di-tert-butylpyridine, 97%
2,6-Di-tert-butylpyridine, 97%
2,6-Di-tert-butylpyridine, 97%
2,6-Di-tert-butylpyridine, 97%
Thermo Scientific Chemicals
2,6-Di-tert-butylpyridine, 97%
CAS: 585-48-8 | C13H21N | 191.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA204640250
Price (MYR)
3,265.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
3,265.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 6-Di-tert-butylpyridine
Name Note97%
CAS585-48-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning