Androsterone, 97%
Androsterone, 97%
Androsterone, 97%
Thermo Scientific Chemicals
Androsterone, 97%
CAS: 53-41-8 | C19H30O2 | 290.447 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA204190010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAndrosterone
Name Note97%
CAS53-41-8
ColorWhite to Beige
RTECS NumberBV8053000