Dowex™ 50WX4, 200-400 mesh, ion-exchange resin
Dowex™ 50WX4, 200-400 mesh, ion-exchange resin
Dowex™ 50WX4, 200-400 mesh, ion-exchange resin
Dowex™ 50WX4, 200-400 mesh, ion-exchange resin
Dowex™ 50WX4, 200-400 mesh, ion-exchange resin
CAS: 11113-61-4
Have Questions?
Catalog NumberQuantityPackaging
FSA203055000500 gPlastic Bottle
Catalog number FSA203055000
Price (MYR)
-
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialDowex™ 50WX4
Physical FormBeads
CAS11113-61-4
Particle SizeParticle size analysis: on 100 mesh (cumulative): <15%, through 325 mesh: <15%
PackagingPlastic Bottle