Tolylene 2,4-diisocyanate, 75%, technical
Tolylene 2,4-diisocyanate, 75%, technical
Tolylene 2,4-diisocyanate, 75%, technical
Tolylene 2,4-diisocyanate, 75%, technical
Thermo Scientific Chemicals
Tolylene 2,4-diisocyanate, 75%, technical
CAS: 584-84-9 | C9H6N2O2 | 174.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA202502500
Price (MYR)
389.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
389.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTolylene 2,4-diisocyanate
CAS584-84-9
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Fatal if inhaled.
May cause an allergic skin reaction.
Suspected of cau
Health Hazard 3GHS P Statement
Immediately call a POISON CENTER or doctor/physician.
If skin irritation occurs: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
If skin irritat