Tannic acid, 95%
Tannic acid, 95%
Tannic acid, 95%
Thermo Scientific Chemicals
Tannic acid, 95%
CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
1 kg
Packaging:
View all
Plastic bottle
Catalog number FSA202425000
Price (MYR)
534.00
EA
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
534.00
EA
Add to cart
Specifications
Chemical Name or MaterialTannic acid
CAS1401-55-4
Melting Point218.0°C
ColorWhite to Yellow
Assay Percent Range95%