Amberlyst™ A26, OH-form, ion-exchange resin
Amberlyst™ A26, OH-form, ion-exchange resin
Amberlyst™ A26, OH-form, ion-exchange resin
Amberlyst™ A26, OH-form, ion-exchange resin
Thermo Scientific Chemicals
Amberlyst™ A26, OH-form, ion-exchange resin
CAS: 39339-85-0
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Plastic bottle
Catalog number FSA202180010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlyst™ A26
CAS39339-85-0
PackagingPlastic bottle
MDL NumberMFCD00145579
Density0.6750g/mL
View more
Ion exchange resin
RUO – Research Use Only