Amberlite™ CG-50 - type 1, ion-exchange resin
Amberlite™ CG-50 - type 1, ion-exchange resin
Amberlite™ CG-50 - type 1, ion-exchange resin
Amberlite™ CG-50 - type 1, ion-exchange resin
Thermo Scientific Chemicals
Amberlite™ CG-50 - type 1, ion-exchange resin
CAS: 9042-11-9
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA202111000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAmberlite™ CG-50 - type 1
CAS9042-11-9
Recommended StorageIndoors +20°C
PackagingPlastic bottle
MDL NumberMFCD00132693
View more
Ion exchange resin
RUO – Research Use Only