Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
Thermo Scientific Chemicals
Tetrabutylammonium fluoride, 1M solution in THF, containing ca. 5% water
CAS: 429-41-4 | C16H36FN | 261.47 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number FSA201955000
Price (MYR)
1,589.00
BOT
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (MYR)
1,589.00
BOT
Add to cart
Chemical Identifiers
CAS429-41-4, 109-99-9, 7732-18-5
Specifications
Chemical Name or MaterialTetrabutylammonium fluoride
Name Note1M Solution in THF, Containing ca 5% Water
CAS429-41-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement
Causes severe skin burns and eye damage. Suspected of causing cancer. May cause respiratory irritation. Highly flammable liquid and vapor. May form explosive peroxides. May cause drowsiness or dizziness.
View more
C-X bond formation reagent
RUO – Research Use Only