Sodium borohydride, 98%, pellets (1-2 cm)
Sodium borohydride, 98%, pellets (1-2 cm)
Sodium borohydride, 98%, pellets (1-2 cm)
Sodium borohydride, 98%, pellets (1-2 cm)
Thermo Scientific Chemicals
Sodium borohydride, 98%, pellets (1-2 cm)
CAS: 16940-66-2 | BH4Na | 37.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic Bottle
Catalog number FSA201485000
Price (MYR)
1,128.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
1,128.00
BOT
Add to cart
Specifications
Chemical Name or MaterialSodium borohydride
Melting Point360.0°C
Name Note7/16 in., 98%
CAS16940-66-2
ColorWhite
View more
Reduction reagent
RUO – Research Use Only