Selenium(IV) oxide, 99.8%, (trace metal basis)
Selenium(IV) oxide, 99.8%, (trace metal basis)
Selenium(IV) oxide, 99.8%, (trace metal basis)
Selenium(IV) oxide, 99.8%, (trace metal basis)
Thermo Scientific Chemicals
Selenium(IV) oxide, 99.8%, (trace metal basis)
CAS: 7446-08-4 | O2Se | 110.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
10 g
100 g
Packaging:
Glass bottle
Catalog number FSA200005000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialSelenium(IV) oxide
Melting Point315.0°C
CAS7446-08-4
ColorWhite to Beige
Physical FormPowder