Selenium, 99.5+%, powder, -200 mesh
Selenium, 99.5+%, powder, -200 mesh
Selenium, 99.5+%, powder, -200 mesh
Selenium, 99.5+%, powder, -200 mesh
Thermo Scientific Chemicals
Selenium, 99.5+%, powder, -200 mesh
CAS: 7782-49-2 | Se | 78.97 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
1 kg
Packaging:
Glass bottle
Catalog number FSA198072500
Price (MYR)
990.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
990.00
BOT
Add to cart
Specifications
Boiling Point685.0°C
Chemical Name or MaterialSelenium
Melting Point217.0°C
CAS7782-49-2
ColorColorless to Yellow